TRITON™ X-100

Leverandør: G Bioscienes
Danger

Synonymer: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

DG007
DG007EA 4350 NOK
DG007
TRITON™ X-100
TRITON™ X-100
Kokepunkt: ∼200 °C (1013 hPa)
Smeltepunkt: 6
Tetthet: 1.06 g/cm³ (20 °C)
Flash punkt: 251 °C (closed cup)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00132505
CAS-nummer: 9002-93-1
FN: 3082
ADR: 9 III
Merck Indeks: 13,06793

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR