Optical plate
Leverandør: DIDALAB
POD010240
DIDAPOD010240EA 0 NOK
DIDAPOD010240
Optical plate
Lærings materiell Læremateriale for fysikk Optics Learning Activities
Optical glass components are manufactured with high accuracy polishing and flatness tolerance.
  • Optical polishing
  • Two sides with parallelism less than 1 µm