Diode laser, green, Class II and IIIa
Leverandør: DIDALAB
POD013132
DIDAPOD013132EA 4850 NOK
DIDAPOD013132
Diode laser, green, Class II and IIIa
Lærings materiell Læremateriale for fysikk Optics Learning Activities
Extremely luminous beam, ideal for classroom experiments. Class IIIa (5 mW Power)
  • Highly miniaturised system with glass optic components
Leveringsinformasjon: Provided with Ø 10 mm rod, mains power supply & adapter for use in Class II.