Diode laser, red
Leverandør: DIDALAB
POD013135
DIDAPOD013135EA 0 NOK
DIDAPOD013135
Diode laser, red
Lærings materiell Læremateriale for fysikk Optics Learning Activities
The diameter of the beam is adjustable from 1 to 12 mm.
Leveringsinformasjon: Supplied with Ø 10-mm rod and mains adapter.