Laurent polarimeters
Leverandør: DIDALAB
POD068570 POD068570_UK
DIDAPOD068570EA 14525 NOK
DIDAPOD068570 DIDAPOD068570_UK
Laurent polarimeters
Polarimetere
The Laurent polarimeter is composed of one lens fitted on graduated rule, and one housing for observation tubes, within which the substances to be analysed are introduced.
Designed for measurement of the polarisation direction in light through an optical medium.
Leveringsinformasjon: Supplied with two observation tubes (100 and 200 mm)