Radtape™ labels
Leverandør: DIVERSIFIED BIOTECH
RAD-10
DIVERAD-10EA 0 NOK
DIVERAD-10
Radtape™ labels
Etiketter
These RadTape™ labels are used to identify autograms. They are phosphorescent, non-radioactive adhesive labels that may be cut to the specific size and shape needed for each individual membrane, dried gel or microarray.
  • Ideal for chemilluminescence and radiological autography
  • Great for documentation and archiving
  • Use your own codes and symbols
  • Fast and easy marking system
An ordinary pen may be used to mark the label using the investigator’s own codes and symbols. The custom label may then be peeled from its backing and affixed to the membrane, gel orMore Product Information