Heavy Duty Non Fuming Acidic Descaler, 20 L, Pascal VA5
Leverandør: Diversey
5600001
DIVY5600001EA 890 NOK
DIVY5600001
Heavy Duty Non Fuming Acidic Descaler, 20 L, Pascal VA5
Rengjøringsløsninger Detergenter og overflatevasking
Pascal is a low foaming, heavy duty, non-fuming nitric acid based descaler for use in a wide range of CIP applications in the food and beverage industry.
  • Pascal is highly effective at removing inorganic scale deposits, including calcium oxalate (beer stone)
  • Pascal is low foaming and suitable for use in CIP applications under conditions of high pressure and turbulence
  • Pascal can also be used for open plant cleaning
  • Pascal is highly economical at in use concentrations
  • Pascal is formulated to be non-fuming
  • Pascal is a conductive liquid detergent and suitable