Detergent, concentrated, Shureclean Plus VK9
Leverandør: Diversey
7509854
DIVY7509854EA 1000 NOK
DIVY7509854
Detergent, concentrated, Shureclean Plus VK9
Rengjøringsløsninger Detergenter og overflatevasking
Shureclean Plus VK9 is a concentrated, neutral, high foaming liquid detergent designed for general purpose applications in the food, beverage and dairy industries. This detergent is recommended for manual and soak cleaning of all food preparation equipment, cooking vessels and utensils. It can also be used for cleaning floors, walls and work surfaces.
  • Clear, pale brown liquid
  • Relative density at 20 °C - 1,03
  • Suitable for soft or hard water and pH neutral
  • Non perfumed product