TASKI® Jontec Best Neutral Floor Degreaser, Low Foam
Leverandør: Diversey
7512310 7512309
DIVY7512310EA 570 NOK
DIVY7512310 DIVY7512309
TASKI® Jontec Best Neutral Floor Degreaser, Low Foam
Rengjøringsløsninger Detergenter og overflatevasking
Heavy duty, alkaline floor cleaner. Cleans heavy soil, dirt and grease off all water resistant, non protected hard floors. Especially suitable for heavy duty ceramic and porous tiles cleaning.
  • pH-neutral
  • Good emulsifying power
  • Fast acting, low foaming
  • Versatile application
  • Environmentally responsible for waste water disposal
  • Unique, patented Odour Neutralizing Technology (O.N.T.)
Safe to use on all water resistant hard floors. Ideal for scrubber driers. Removes quickly and thoroughly mineral/synthetic oils and grease, forklift tire marks as well asMore Product Information