TASKI Sani Antikalk Heavy Duty Washroom Cleaners, W3e
Leverandør: Diversey
7513263
DIVY7513263EA 300 NOK
DIVY7513263
TASKI Sani Antikalk Heavy Duty Washroom Cleaners, W3e
Rengjøringsløsninger Detergenter og overflatevasking
Acid-based sanitary cleaner and limescale remover for acid resistant hard surfaces.
  • Sulphamic acid based
  • Contains a corrosions inhibitor
  • Strong descaling properties
  • Unique, patented Odour Neutralizing Technology (O.N.T.)
Effective removal of stubborn calcium/limescale and of common sanitary soil, e.g. body fat and soap residues. Perfectly fit for efficient full surface descaling of floors, walls and other washroomsurfaces. Ready to use, streak free cleaning. Leaves a pleasant, fresh fragrance.
Spray method:
- Spray the solution on a damp cloth andMore Product Information