TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Leverandør: Diversey
7515243
DIVY7515243EA 680 NOK
DIVY7515243
TASKI Sani Calc Pur-Eco W3b Washroom Descaler
Rengjøringsløsninger Detergenter og overflatevasking
Powerful limescale remover for acid resistant hard surfaces.
  • Fast acting acidic, based on citric acid formula
  • Excellent cleaning performance
  • Strong descaling properties
  • No scrubbing necessary at high concentration
Sertifiseringer: EU Ecolabel (EU Flower).