Detergent, Diversey Attack Plus E9e
Leverandør: Diversey
7516886
DIVY7516886EA 1250 NOK
DIVY7516886
Detergent, Diversey Attack Plus E9e
Rengjøringsløsninger Detergenter og overflatevasking
Clear yellow, heavy-duty cleaner containing a mixture of organic solvents and compounds.
  • Good emulsifying formula
  • Removes grease and dirt
  • Medium foaming alkaline formula, pH of neat solution: 12