Vacuum aspiration system, AsPROvac
Leverandør: DON WHITLEY SCIENTIFIC
A07950
DOWIA07950EA 56900 NOK
DOWIA07950
Vacuum aspiration system, AsPROvac
Pumper Vakuum remdrevet pumper
This AsPROvac is an all-in-one laboratory aspirator to ensure biological liquid waste can be aspirated and disposed of quickly and safely.
  • Self-closure connectors avoid the escape of liquids and aerosols
  • Liquid level 'full' indicator
  • Features a hydrophobic filter for user protection
  • All parts that come into contact with the liquid waste are autoclavable
  • Simple, straightforward operation with little training required
  • Lightweight so can be moved around the laboratory with ease
  • Additional ergomomic hand holds on the base for easy lifting and handling of the