Safety Cabinets, Type 90, with Wing Door Technology, CLASSIC LINE PURE
Leverandør: DUPERTHAL
29-200667-006 29-201267-006
DUEH29-200667-006EA 0 NOK
DUEH29-200667-006 DUEH29-201267-006
Safety Cabinets, Type 90, with Wing Door Technology, CLASSIC LINE PURE
Møbler Kjemikalieskap
Classical door technology (door handle), door hung right for the storage of flammable liquids.
  • Outer carcass sheet steel, silver
  • Door sheet steel, silver
  • Inner carcass, stainless steel 1.4301, bottom tray, stainless steel 1.4301
  • Silver similar to RAL 9006
  • Door closes automatically in case of fire, door remains open in every position
  • Cylinder lock for safe access
  • 4× storage shelves, stainless steel 1.4301
  • Exhaust air connection socket on the cabinet roof
Visual inspection of the ventilation opening. Earthing connection on the cabinet roof. interiorMore Product Information