Bench mats for cleanrooms
Leverandør: DYCEM
CZ06BM60045BK CZ06BM60120GR
DYCECZ06BM60045BKEA 5875 NOK
DYCECZ06BM60045BK 115-0516
Bench mats for cleanrooms
Benkepapir Benkepapir
This flexible bench mat can be placed on all kind of surfaces. The closed cell structure prevents the movement and loss of small components used in laboratories to stabilise fragile equipment. Approved for use in all classes of cleanrooms, including microelectronics, micro mechanics, semi-conductor, pharmaceutical, biotech and medical devices.
  • Non-slip and anti-static properties
  • Good shock absorption and stabilisation of sensitive equipment
  • Improves accuracy, precision and assembly time
  • Non-particulating: Helmke drum test
  • Durable and easy to clean
  • Portable,
More Product Information
Bestill nå
SPESIFIKASJONER
Farge Grå