Ethyl-beta-D-glucuronide, certified reference material, Cerilliant®, Supelco®
Leverandør: Merck
E-015-1ML
E-015-1MLEA 3300 NOK
E-015-1ML
Ethyl-beta-D-glucuronide, certified reference material, Cerilliant®, Supelco®
Standarder Organiske standarder
Ethyl ß-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alchohol consumption.
This Snap-N-Spike® Reference Solution is applicable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing.
Forpakning: Snap-N-Spike®/Snap-N-Shoot®.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e8e1e0f3bb41cf2f0c177501e89c088ca3cb994&vid=408fdfc5b59df7ae1daa038cc9319380a7c69910&prid=web&clid=SAPDAM