VWR®, TE (Tris-EDTA) buffer, pH 8,0 for biotechnology

Leverandør: VWR Chemicals

E112-100MLEA 660 NOK
E112-100ML E112-500ML
VWR®, TE (Tris-EDTA) buffer, pH 8,0 for biotechnology
Buffere Buffere, kloning

Ready to use Tris-EDTA buffer.


  • Composition: 10 mM Tris, 1 mM EDTA
  • pH 8,0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR