VWR®, Aprotinin
Leverandør: VWR Chemicals
E429-50MGEA 9325 NOK
E429-50MG
VWR®, Aprotinin
Proteiner
A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin. Not an inhibitor of Factor Xa and thrombin.
Activity (KIU/mg)      4500