VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Leverandør: VWR Chemicals
E504-500MLEA 580 NOK
E504-500ML
VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Buffere Buffere, cellekultur
137 mM sodium chloride, 2,7 mM potassium chloride, 12 mM phosphate buffer
  • pH 7,4