VWR® Sodium Acetate Buffer 3,0 M Solution, pH 5,2
Leverandør: VWR Chemicals
E521-100MLEA 2525 NOK
E521-100ML
VWR® Sodium Acetate Buffer 3,0 M Solution, pH 5,2
Buffere Buffere, kloning
With diethyl pyrocarbonate (DEPC).
  • DNase-free
  • RNase-free