Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology

Leverandør: VWR Chemicals

E529-500MLEA 905 NOK
E529-500ML
Sodium chloride 5 mol/l (5 N) in aqueous solution for biotechnology
Natriumklorid

Common source of salts for media propagation of many organisms.

Formel: NaCl
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5
EINECS: 231-598-3

Bestill nå

Spesifikasjon Testresultater

Conductivity (1 %, Water) 5000 - 7000 umhos
Identification PASS
pH @25 °C 5.0 - 9.0

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR