Bradford's reagent, Biotechnology Grade for quantitative protein determination, for quantitative protein determination
Leverandør: VWR Chemicals
E530-1LEA 5875 NOK
E530-1L
Bradford's reagent, Biotechnology Grade for quantitative protein determination, for quantitative protein determination
Bradford's reagent
Bestill nå
Spesifikasjon Testresultater
Abs.at peak A 0.95 - 1.6
Abs. at peak A/Abs. at 595 nm >1.7
Bradford protein assay Pass
Characteristic pattern Pass
Peak A 463 nm - 467 nm
pH (at 25 °C) 0.5 - 1.5