VWR®, dTTP solution
Leverandør: VWR Chemicals
E542-40UMEA 3425 NOK
E542-40UM
VWR®, dTTP solution
Nukleinsyre reagenser Nukleotider
Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA.
  • Building blocks for the amplification of DNA
  • High purity grade
DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule.