Estriol 3-sulphate sodium salt ≥98% (by TLC), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning


M-Clarity™ quality level = MQ200

E6375-10MG E6375-25MG
E6375-10MGEA 4710 NOK
E6375-10MG E6375-25MG
Estriol 3-sulphate sodium salt ≥98% (by TLC), Sigma-Aldrich®
Estriol 3-sulphate sodium salt

Estriol 3-sulfate sodium salt, (1,3,5(10)-Estratriene-3,16alpha,17beta-triol 3-sulfate; 3,16alpha,17beta-Trihydroxy-1,3,5(10)-estratriene 3-sulfate)


Packaging: Package size based on steroid content.

Formula: C₁₈H₂₃NaO₆S
Storage Temperature: Room Temperature
MDL Number: MFCD00056547
CAS Number: 5150-64-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=176c0e8b082d6165a809de31b7e5a05d617b1a36&vid=d18b11954a74b3a81c20f40805473558980c6a5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Form) Powder or Lyophilised Powder
Appearance (Colour) White to Off-White
Solubility (Solvent) Methanol
Solubility (Conc) 19.60 - 20.40 mg/ml
Solubility (Turbidity) Clear
Solubility (Colour) Colorless to Faint Yellow
Purity (TLC) ≥98.00 %
Stabiliser Content(Stabiliser) Methyl Glucamine
Stabiliser Content (Method) UV
Stabiliser Content (%) 25.00 - 35.00 %
Storage Conditions Under Argon
Recommended Retest Period 5 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR