Fast green FCF, Certifiable Grade

Leverandør: VWR Chemicals
Warning

Synonymer: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt

E772-25GEA 7350 NOK
E772-25G
Fast green FCF, Certifiable Grade
Fast green FCF

Bluish-green dye that is soluble in alcohol. Can be used with safranin as a stain for plant tissue and for demonstrating nuclear histones.

Formel: C₃₇H₃₄N₂Na₂O₁₀S₃
MW: 808.86 g/mol
Smeltepunkt: 290
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013053
CAS-nummer: 2353-45-9
EINECS: 219-091-5
Merck Indeks: 13,03970

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR