Winlog.pro Evaluation Software for Ebro Dataloggers
Leverandør: EBRO
1340-2450
EBRO1340-2450EA 17375 NOK
EBRO1340-2450
Winlog.pro Evaluation Software for Ebro Dataloggers
Programvare Laboratoriesoftware
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For professional software.
  • Contains all properties of Winlog.basic
  • Enables real-time monitoring with radio wave data loggers
  • Current version: 3.1
Supports: EBI 12, EBI 10, EBI 11, EBI 20, EBI 40, EBI 100, EBI 300, EBI 310, EBI 330. Integration of images and diagrams into reports possible. FDA 21 CFR Part 11 functionality also possible. Optional IQ-/OQ documentation also available. Formula editor forMore Product Information