Winlog.Web Server Based Monitoring Software for EBI 25
Leverandør: EBRO
1340-2490 1340-2491
EBRO1340-2490EA 18200 NOK
EBRO1340-2490 EBRO1340-2491
Winlog.Web Server Based Monitoring Software for EBI 25
Programvare Laboratoriesoftware
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. Supports: data loggers EBI 25 series, MODBUS IP devices. Professional version for multiple users.
  • Connection of the interface IF 400 via USB and ethernet
  • FDA 21 CFR Part 11 functionality
  • Administration of large quantities of data
  • IQ-/OQ documentation available
  • Current version: 3.11
Web-based server solution: the measurement data can be read on all computers and smartphones which are connected with theMore Product Information