Robust and Compact High Temperature Data Logger, EBI 11-T210
Leverandør: EBRO
1340-6260
EBRO1340-6260EA 19500 NOK
EBRO1340-6260
Robust and Compact High Temperature Data Logger, EBI 11-T210
Dataloggere
The EBI 11-T210 temperature data logger is ideal for versatile routine checks in the food and pharmaceutical industries. With an extended measuring range (–30 to +140 °C), high–quality PT1000 sensor, very small size that allows it to fit easily into most bottles or small packaging, robust IP 68 protection class, and the option of use in autoclaves, it ensures reliable pasteurisation control and food safety.
  • Rigid, external temperature sensor: Ø 3 mm, 20 mm long
  • Waterproof and vapour-tight
  • Compact size
This data logger was developed for routine checksMore Product Information