Basic Temperature Monitoring Kit for Versatile Applications, SL4211 Set: EBI 12-T101
Leverandør: EBRO
1340-6601
EBRO1340-6601EA 28475 NOK
EBRO1340-6601
Basic Temperature Monitoring Kit for Versatile Applications, SL4211 Set: EBI 12-T101
Dataloggere
The SL4211 set by ebro, featuring EBI 12-T101 temperature loggers, is a versatile solution for basic temperature monitoring in industrial, food, and laboratory applications, offering reliable and accurate data collection for various processes.
  • Basic temperature monitoring set for diverse applications
  • Includes EBI 12-T101 temperature data logger
  • High-precision monitoring
  • Suitable for industrial, food production, and laboratory environments
  • Compact and durable instruments with high IP protection
  • Factory calibration certificate
The SL4211 set by ebroMore Product Information
Bestill nå
SPESIFIKASJONER
Modell EBI 12-T101
Environmentally Preferable
Sensor PT1000, class A
Grensesnitt
Beskyttelsesklasse IP 68
Registreringsintervaller 1 s - 24 h
Vekt ca. 90 g
Dataminne - Antall avlesninger 27840
Batterier Lithium battery, 3,6 V