Temperature Data Logger for Process Validation in Steam Sterilisation, EBI 12-T690
Leverandør: EBRO
1340-6635
EBRO1340-6635EA 0 NOK
EBRO1340-6635
Temperature Data Logger for Process Validation in Steam Sterilisation, EBI 12-T690
Dataloggere
The EBI 12-T690 is a high-precision temperature data logger designed for process validation in steam sterilisers and batch control in production. With a temperature range of –20 to +150 °C and an accuracy of up to ±0,05 °C, it provides reliable data for critical monitoring processes.
  • Suitable for steam sterilisers, process validation
  • Flexible cable probe, 1,20 mm
  • Factory calibration certificate at 0, 60, and 134 °C
The EBI 12-T690 temperature data logger by ebro is ideal for demanding applications like process validation in steam sterilisers and batchMore Product Information