Hightextra® Smooth Wipes
Leverandør: ECOLINE NV SA
HTS0060R05300W
ECOLHTS0060R05300WEA 1225 NOK
ECOLHTS0060R05300W
Hightextra® Smooth Wipes
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
The combination of woodpulp and continuous polyester fibres creates a super strong wipe, even when used wet, that copes with tougher cleaning too.