Streptomycin sulphate

Leverandør: EDQM
Warning

S1400000
EDQMS1400000EA 3225 NOK
EDQMS1400000
Streptomycin sulphate
Streptomycin sulphate
Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Smeltepunkt: 250 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00037023
CAS-nummer: 3810-74-0
EINECS: 223-286-0
FN: 2811
ADR: 6.1,III
Merck Indeks: 12,08983

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR