Organic reference standards, rivastigmine, Ph. Eur., RS

Leverandør: EDQM

Y0001516 Y0001515 Y0001485
EDQMY0001516EA 3225 NOK
EDQMY0001516 EDQMY0001515 EDQMY0001485
Organic reference standards, rivastigmine, Ph. Eur., RS
Standarder Organiske standarder

Bestill nå

SPESIFIKASJONER

Beskrivelse Rivastigmine hydrogen tartrate Rivastigmine impurity D, 3-[(1R)-1-(dimethylamino)ethyl]phenyl ethyl(methyl)carbamate ((R)-enantiomer) Rivastigmine for system suitability
CAS-nr 129101-54-8 415973-05-6 129101-54-8
Formel C₁₈H₂₈N₂O₈ C₁₄H₂₂N₂O₂ C₁₈H₂₈N₂O₈

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR