Digital multimeter, autoranging with non contact voltage detector, MS8360F
Leverandør: EDUCATIONSCIENCE COLLECTION
MS8360F
EDSCMS8360FEA 0 NOK
EDSCMS8360F
Digital multimeter, autoranging with non contact voltage detector, MS8360F
Multimetre Multimeter, digital
Multimeter to test DC voltage, AC voltage, DC current, AC current, resistance, capacitance, frequency and duty cycle.