Kits: restriction enzyme cleavage patterns of DNA, Ready-to-Load™
Leverandør: EDVOTEK
102 102-C
EDVO102EA 1400 NOK
EDVO102 EDVO102-C
Kits: restriction enzyme cleavage patterns of DNA, Ready-to-Load™
Lærings materiell Læremateriale for biologi Genetics Learning Activities
Plasmid and lambda DNA are pre-digested with restriction endonucleases that recognise and cut double-stranded DNA within or near defined base sequences. Digests are separated by agarose gel electrophoresis.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Leveringsinformasjon: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNAMore Product Information