Experimentation kits, molecular weight determination of proteins (agarose-based)
Leverandør: EDVOTEK
110
EDVO110EA 1775 NOK
EDVO110
Experimentation kits, molecular weight determination of proteins (agarose-based)
Lærings materiell Læremateriale for biologi Genetics Learning Activities
This experiment introduces a simple method to determine protein subunit molecular weights using horizontal electrophoresis. As the protein standards and 'unknowns' are prestained, the separation of proteins can be observed during electrophoresis. Included in the experiment is protein-grade agarose, which provides an alternative to the use of polyacrylamide gels.
  • For 4 lab groups
  • Complete in 1 hour followed by overnight staining
Also required: Horizontal electrophoresis apparatus, power supply, white light visualisation system, 5 to 50 µl adjustable orMore Product Information