Nucleic acid testing for COVID-19
Leverandør: EDVOTEK
123
EDVO123EA 1400 NOK
EDVO123
Nucleic acid testing for COVID-19
Lærings materiell Læremateriale for biologi Genetics Learning Activities
Hands-on simulation of molecular testing for COVID-19.
  • Students learn about characteristics of Coronaviruses
  • Includes all required reagents
  • Use DNA Electrophoresis to analyse simulated samples
  • Activity can be completed in 45 minutes
SARS-CoV-2 is a novel coronavirus that has caused a worldwide outbreak of respiratory disease beginning in 2019.
In this simulated medical test, we will use electrophoresis to detect the presence of the SARS-CoV-2 virus in samples from patients with symptoms of COVID-19.
Additional items needed but not included: DNAMore Product Information