Experimentation kits, blood-based cancer diagnostics
Leverandør: EDVOTEK
141
EDVO141EA 1475 NOK
EDVO141
Experimentation kits, blood-based cancer diagnostics
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
Cancer cells differ from normal cells by the combinations of proteins that are present on their surfaces. Antibodies against these proteins will specifically bind to cancer cells and not to normal cells. This allows early detection of cancer and potentially a way of delivering cancer therapies. In this simulation experiment the reaction of cancer cell markers and their corresponding antigens are demonstrated.
  • For 10 lab groups
  • Complete in 35 minutes
Leveringsinformasjon: Kit includes instructions, microtitre plates, cancer cell markers, normal cellMore Product Information