Experimentation kits, determination of protein molecular weight (polyacrylamide-based)
Leverandør: EDVOTEK
153
EDVO153EA 1125 NOK
EDVO153
Experimentation kits, determination of protein molecular weight (polyacrylamide-based)
Lærings materiell Læremateriale for biologi Genetics Learning Activities
Using prestained LyphoProteins, subunit molecular weights are determined by analysis using denaturing SDS vertical polyacrylamide gel electrophoresis. Prestained proteins with unknown molecular weights are assigned molecular weights based on the relative mobility of prestained standard protein markers.
  • For 6 lab groups (sharing 3 gels)
  • Electrophoresis 60 minutes, staining 20 minutes, destaining 2 hours
Also required: 3 polyacrylamide gels (12%), vertical gel electrophoresis apparatus, power supply, white light box, 5 to 50 µl adjustable or 20 µl fixedMore Product Information