Experimentation kits, detection of a simulated infectious agent
Leverandør: EDVOTEK
166
EDVO166EA 1775 NOK
EDVO166
Experimentation kits, detection of a simulated infectious agent
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
An infectious outbreak requires prompt and accurate identification of the biological agent. Often, early clinical symptoms are first identified in exposed individuals and then infectious agents are identified by lab tests. In this experiment, students transmit a simulated infectious agent (chemical dye) between classmates which is only visible under long UV light. The pattern of transmission and primary source is documented.
  • For 25 students
  • Requires 30 to 45 minutes
Leveringsinformasjon: Kit includes instructions, reagents for simulating an infectiousMore Product Information