Experimentation kits, DNA fingerprinting using restriction enzymes
Leverandør: EDVOTEK
225
EDVO225EA 1975 NOK
EDVO225
Experimentation kits, DNA fingerprinting using restriction enzymes
Lærings materiell Læremateriale for biologi Biochemistry Learning Activities
This experiment helps teach students about restriction enzyme digests in the context of forensic science. Students cut DNA with restriction enzymes and then compare the banding pattern of the crime scene DNA versus that of two suspects using agarose gel electrophoresis.
  • For 6 lab groups
  • Complete in 90 minutes
Also required: Electrophoresis tank and power supply, 5 to 50 µl adjustable or 5, 10, 15 and 40 µl fixed volume micropipettes, water bath, balance, and white light box.
Leveringsinformasjon: Kit includes instructions, "crime scene" Ready-to-More Product Information