Experimentation kits, introduction to ELISA reactions
Leverandør: EDVOTEK
269
EDVO269EA 1975 NOK
EDVO269
Experimentation kits, introduction to ELISA reactions
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
Students learn the basic principles of the Enzyme-linked Immunosorbent Assay (ELISA) using this precise and sensitive antibody-based detection kit.
  • For 10 Lab Groups
  • Complete in 45 minutes
Also required: Distilled or deionised water, 37 °C incubator, automatic micropipettes with tips, and laboratory glassware.
Leveringsinformasjon: Kit includes instructions, antigens, primary and secondary antibodies, peroxide co-substrate, hydrogen peroxide, ABTS substrate, phosphate buffered saline, tubes, plates, and transfer pipettes.