Experimentation kits, antigen-antibody interaction: the ouchterlony procedure
Leverandør: EDVOTEK
270
EDVO270EA 1975 NOK
EDVO270
Experimentation kits, antigen-antibody interaction: the ouchterlony procedure
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
This experiment introduces students to the principles of antigen-antibody interactions by using the Ouchterlony procedure. Antibodies and antigens form complexes that precipitate, making it possible to assay antibody-antigen systems. The binding interaction results in the formation of a white precipitate after diffusion in agarose.
  • For 10 Lab Groups
  • Complete in 35 minutes followed by overnight incubation
Also required: Automatic micropipettes with tips, 5 or 10 ml pipettes, 55 °C water bath, measuring spatulas or toothpicks, microwave or hot plate,More Product Information