Experimentation kits, in search of the "Kissing Disease"
Leverandør: EDVOTEK
274
EDVO274EA 1975 NOK
EDVO274
Experimentation kits, in search of the "Kissing Disease"
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
Infectious mononucleosis is commonly known as the 'kissing disease'. The causative agent is Epstein-Barr virus (EBV), which can be transmitted through saliva during kissing. In this experiment, students search for the presence of EBV using the ELISA reaction to detect specific viral proteins.
  • For 10 lab groups
  • Requires 50 minutes
Also required: 37 °C incubator, automatic micropipettes with tips.
Leveringsinformasjon: Kit includes instructions, samples, antigens and antibodies, various solutions and reagents, pipettes and microtest tubes.