Experimentation kits, simulation of HIV detection by western blot (agarose-based)
Leverandør: EDVOTEK
275
EDVO275EA 1975 NOK
EDVO275
Experimentation kits, simulation of HIV detection by western blot (agarose-based)
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
The second assay used to confirm a positive HIV ELISA result is the Western blot. Using this kit, students separate protein samples from hypothetical patients on agarose gels. The proteins are then transferred to a membrane and simulated HIV proteins are detected.
  • For 6 lab groups
  • Electrophoresis 45 minutes; blot overnight; detection 25 minutes
Also required: Electrophoresis tank, power supply, isopropanol, glacial acetic acid, and 37 °C incubator.
Leveringsinformasjon: Kit includes instructions, positive and negative controls, simulated patientMore Product Information