Experimentation kits, exploring biotechnology with green fluorescent protein (GFP)
Leverandør: EDVOTEK
303
EDVO303EA 2800 NOK
EDVO303
Experimentation kits, exploring biotechnology with green fluorescent protein (GFP)
Lærings materiell Læremateriale for biologi Biochemistry Learning Activities
Four experimental modules are combined into one experiment to provide a comprehensive biotechnology exploration focusing on the green fluorescent protein (GFP). Bacterial cells are transformed to express the GFP. The transformed cells are then grown and the GFP purified by column chromatography. Finally, the purity of the protein fractions are analysed by SDS polyacrylamide electrophoresis.
  • For 6 lab groups
  • Transformation 50 minutes; overnight incubation; chromatography 45 minutes; optional electrophoresis 90 minutes
Also required: Incubator, waterMore Product Information