Experimentation kits, mitochondrial DNA analysis using PCR
Leverandør: EDVOTEK
332
EDVO332EA 3825 NOK
EDVO332
Experimentation kits, mitochondrial DNA analysis using PCR
Lærings materiell Læremateriale for biologi PCR Learning Activities
The mitochondria are thought to have evolved from a symbiotic relationship between prokaryotic and eukaryotic cells. Mitochondria have their own DNA and are only inherited via the maternal line. In this experiment, students amplify two regions of their mitochondrial DNA.
  • For 25 students
  • Set up 30 minutes; PCR 2 hours or overnight; electrophoresis 45 minutes
Also required: Micropipettes (5 to 50 µl) with tips, water bath, thermal cycler, electrophoresis tank and power supply.
Leveringsinformasjon: Kit includes instructions, proteinase K, PCR beads,More Product Information