Experimentation kits, introduction to plant cell culture
Leverandør: EDVOTEK
908
EDVO908EA 1900 NOK
EDVO908
Experimentation kits, introduction to plant cell culture
Lærings materiell Læremateriale for biologi Plant Learning Activities
Genetic modification of plants is a controversial area of biotechnology. All such experiments in plants begin with establishing plant cells in culture. This involves de-differentiating plant cells to form plant 'stem cells'. In this experiment, students establish cell cultures of African violets from leaves. They then use plant growth regulators to encourage root growth from the cultured cells, and produce a mature plant.
  • For 10 lab groups
  • Complete in 60 minutes plus several weeks for growth
Also required: Healthy African violet (Saintpaulia ionantha).More Product Information