Experimentation kit, bioremediation by oil eating bacteria
Leverandør: EDVOTEK
956
EDVO956EA 1825 NOK
EDVO956
Experimentation kit, bioremediation by oil eating bacteria
Lærings materiell Læremateriale for geofag Natural Resources Learning Activities
Oil spills cause devastation to the environment, killing sea life, birds, and coastal plants. Spraying areas of contamination with oil-eating microbes accelerates the degradation of the oil. This process is known as bioremediation. In this open-ended experiment, students grow a mixture of oil-eating bacteria and observe their effectiveness at degrading a variety of oils.
  • For 10 lab groups
  • After establishment of cultures, lab requires 50 minutes (can be done over several days or weeks)
Also required: Incubator, growth flasks, vegetable oil (or otherMore Product Information