Experimentation kits, DNA damage and repair
Leverandør: EDVOTEK
957
EDVO957EA 1825 NOK
EDVO957
Experimentation kits, DNA damage and repair
Lærings materiell Læremateriale for biologi Physiology and Health Learning Activities
According to the World Health Organisation, between 2 and 3 million cases of skin cancer occur globally every year. Many of these cases are caused by preventable damage to DNA by UV light. In this experiment, students expose plasmid DNA to shortwave UV light to simulate the effect of sunbathing. The DNA is then analysed by agarose gel electrophoresis to observe the damage.
  • For 10 lab groups
  • Complete in 2 hours
Also required: UV transilluminator, electrophoresis tank and power supply.
Leveringsinformasjon: Kit includes instructions, standard DNAMore Product Information